missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals O-Acetyl-L-carnitinhydrochlorid
Click to view available options
Menge:
5 g
25 g
Beschreibung
O-Acetyl-L-carnitine hydrochloride is used as a cholinergic agonist that stimulates neuronal response to serotonin and acetylcholine. Acetylcarnitine has antinociceptive activity that may be mediated by enhanced activity of muscarinic cholinergic receptors or mGlu2 glutamate receptors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Menge | 5 g |
Beschreibung | Weiß |
Inhalt und Lagerung | 2°bis 8 °C |
Zur Verwendung mit (Anwendung) | Schwacher cholinerger Agonist |
Formelmasse | 239,70g/mol |
Empfindlichkeit | Hygroskopisch |
Produkttyp | Signaltransduktionsreagenz |
Form | Weißes kristallines Pulver |
Beilstein | 4340103 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'