Learn More
OBSL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA557640
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: VRVVAPEAAQ TRVRSTPGGD LELVVHLSGP GGPVRWYKDG ERLASQGRVQ LEQAGARQVL RVQGARSGDA GEYLCDAPQD SRIFLVSVE Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 35%.
OBSL1 Polyclonal antibody specifically detects OBSL1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
OBSL1 | |
Polyclonal | |
Unconjugated | |
OBSL1 | |
KIAA0657; obscurin like 1; obscurin-like 1; obscurin-like protein 1; OBSL1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23363 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75147 | |
OBSL1 | |
Recombinant protein corresponding to Human OBSL1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only