Learn More
Optineurin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA551882
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LELAEKALAS KQLQMDEMKQ TIAKQEEDLE TMTILRAQME VYCSDFHAER AAREKIHEEK EQLALQLAVL LKENDAFEDG GRQSLMEMQS RHGARTSDSD QQAYLVQRGA EDRDWRQQRN IPIHSCPKCG EVL Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 88%.
Optineurin Polyclonal antibody specifically detects Optineurin in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Optineurin | |
Polyclonal | |
Unconjugated | |
OPTN | |
14.7K-interacting protein 2; 4930441O07Rik; Ag9 C5; ALS12; E3-14.7K-interacting protein; FIP 2; FIP2; FIP-2; FIP-2-like protein; GLC1E; HIP 7; HIP7; HIP-7; Huntingtin interacting protein L; huntingtin yeast partner L; Huntingtin-interacting protein 7; huntingtin-interacting protein L; HYPL; NEMO-related protein; NRP; Optic neuropathy-inducing protein; optineurin; Optn; TFIIIA IntP; TFIIIA-IntP; transcription factor IIIA-interacting protein; transcrption factor IIIA-interacting protein; tumor necrosis factor alpha-inducible cellular protein containing leucine zipper domains; Uban | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10133 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96CV9 | |
OPTN | |
Recombinant protein corresponding to Human Optineurin | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only