missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR10AD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA562089
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: NKEIKEAMVR ALGRTRLAQP QSV Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 70%.
OR10AD1 Polyclonal antibody specifically detects OR10AD1 in Human samples. It is validated for Immunohistochemistry.Spezifikation
OR10AD1 | |
Polyclonal | |
Unconjugated | |
OR10AD1 | |
olfactory receptor 10AD1; olfactory receptor family 10 subfamily AD member 1; olfactory receptor OR12-1; olfactory receptor, family 10, subfamily AD, member 1 pseudogene; OR10AD1; OR10AD1P; OR12-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
121275 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NGE0 | |
OR10AD1 | |
Recombinant protein corresponding to Human OR10AD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only