missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR10H5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA548396
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
OR10H5 Polyclonal antibody specifically detects OR10H5 in Human samples. It is validated for Western Blot.Spezifikation
OR10H5 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q8NGA6 | |
OR10H5 | |
KLH conjugated synthetic peptide between 245-273 amino acids from the C-terminal region of human OR10H5. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
OR10H5 | |
Olfactory receptor 10H5; olfactory receptor family 10 subfamily H member 5; olfactory receptor OR19-25; Olfactory receptor OR19-26; olfactory receptor, family 10, subfamily H, member 5; OR10H5; OR19-25; OR19-26 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
284433 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |