Learn More
OR10J3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA560175
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LKPKSQSSLG QDRLISVTYT HHSPTEPCCV QPEEQGGQRC SAQSRGAKNS VSLMKRGCEG FSFAFINMY Highest antigen sequence identity to the following orthologs: Mouse - 29%, Rat - 30%.
OR10J3 Polyclonal antibody specifically detects OR10J3 in Human samples. It is validated for Immunohistochemistry.Spezifikation
OR10J3 | |
Polyclonal | |
Unconjugated | |
OR10J3 | |
Olfactory receptor 10J3; olfactory receptor family 10 subfamily J member 3; olfactory receptor OR1-25 pseudogene; olfactory receptor, family 10, subfamily J, member 3 pseudogene; OR10J3; OR10J3P; OR1-25 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
441911 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5JRS4 | |
OR10J3 | |
Recombinant protein corresponding to Human OR10J3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only