missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR10Z1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563519
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: GQTNVTSWRD FVFLGFSSSG Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 65%.
OR10Z1 Polyclonal antibody specifically detects OR10Z1 in Human samples. It is validated for Immunohistochemistry.Spezifikation
OR10Z1 | |
Polyclonal | |
Unconjugated | |
OR10Z1 | |
Olfactory receptor 10Z1; olfactory receptor family 10 subfamily Z member 1; olfactory receptor OR1-15; olfactory receptor, family 10, subfamily Z, member 1; OR10Z1; OR1-15 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
128368 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NGY1 | |
OR10Z1 | |
Recombinant protein corresponding to Human OR10Z1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only