Learn More
OR4K1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA555999
Additional Details : Netto-Gewicht : 0.02000kg
Description
Immunogen sequence: TVDLPFCGPN EVDSFFCDLP LVIELACMDT YEMEIMTLTN Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 68%.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.Specifications
OR4K1 | |
Polyclonal | |
Unconjugated | |
OR4K1 | |
Olfactory receptor 4K1; olfactory receptor family 4 subfamily K member 1; olfactory receptor OR14-19; olfactory receptor, family 4, subfamily K, member 1; OR14-19; OR4K1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79544 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NGD4 | |
OR4K1 | |
Recombinant protein corresponding to Human OR4K1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only