missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR52I1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA549176
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
OR52I1 Polyclonal antibody specifically detects OR52I1 in Human samples. It is validated for Western Blot.Spezifikation
OR52I1 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q8NGK6 | |
OR52I1 | |
KLH conjugated synthetic peptide between 212-240 amino acids from the C-terminal region of human OR52I1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
OR52I1 | |
Olfactory receptor 52I1; olfactory receptor family 52 subfamily I member 1; olfactory receptor OR11-13; olfactory receptor, family 52, subfamily I, member 1; OR11-13; OR52I1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
390037 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |