missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR5B12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA567657
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.Spezifikation
OR5B12 | |
Polyclonal | |
Unconjugated | |
OR5B12 | |
OR5B12P, OR5B16 | |
Rabbit | |
Affinity chromatography | |
RUO | |
390191 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q96R08 | |
OR5B12 | |
Synthesized peptide derived from human OR5B12. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |