missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR5V1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA561290
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: KDIKEAVKTI GSKWQPPISS LDSKLTY Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 63%.
Odorant receptor.Spezifikation
OR5V1 | |
Polyclonal | |
Unconjugated | |
OR5V1 | |
6M1-21; hs6M1-21; Olfactory receptor 5V1; olfactory receptor family 5 subfamily V member 1; olfactory receptor OR6-26; olfactory receptor, family 5, subfamily V, member 1; OR5V1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81696 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UGF6 | |
OR5V1 | |
Recombinant protein corresponding to Human OR5V1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only