missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR6N2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563334
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: SLRNKEIIKA IKRTIFQKGD KASLAH Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 69%.
Odorant receptor.Spezifikation
OR6N2 | |
Polyclonal | |
Unconjugated | |
OR6N2 | |
Olfactory receptor 6N2; olfactory receptor family 6 subfamily N member 2; olfactory receptor OR1-23; olfactory receptor, family 6, subfamily N, member 2; OR1-23; OR6N2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81442 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NGY6 | |
OR6N2 | |
Recombinant protein corresponding to Human OR6N2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only