Learn More
ORP7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA557512
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: SSENEGSEEE ESCTSEITTS LSEEMLDLRG AERCQKGGCV PGRPMGPPRR RCLPAASGPG ADVSLWNILR NNIGKDLS Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 86%.
This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. Two transcript variants encoding the same isoform have been identified.Spezifikation
ORP7 | |
Polyclonal | |
Unconjugated | |
OSBPL7 | |
OSBPL7; ORP7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114881 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BZF2 | |
OSBPL7 | |
Recombit protein corresponding to Human ORP7 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only