missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human OSCAR Antibody, R&D Systems™
Goat Polyclonal Antibody
N.V.
Spezifikation
Antigen | OSCAR |
---|---|
Konzentration | LYOPH |
Verdünnung | Western Blot 0.1 ug/mL |
Anwendungen | Western Blot |
Klassifikation | Polyclonal |
Beschreibung
OSCAR Polyclonal specifically detects OSCAR in Human samples. It is validated for Western Blot.Spezifikation
OSCAR | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
hOSCAR, MGC33613, osteoclast associated receptor OSCAR-S1, osteoclast associated receptor OSCAR-S2, osteoclast associated, immunoglobulin-like receptor, osteoclast-associated immunoglobulin-like receptor, Osteoclast-associated receptor, PIgR-3, PIGR3, Poly-Ig receptor 3, Polymeric immunoglobulin receptor 3 | |
OSCAR | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
AAM19095 | |
126014 | |
Mouse myeloma cell line NS0-derived recombinant human OSCAR isoform 2 Asp19-Asn229 (Asn158Ser) Accession # AAM19095 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human OSCAR in direct ELISAs and Western blots. In these formats, approximately 25% cross-reactivity with recombinant mouse OSCAR is observed. |
For Research Use Only