Learn More
OTOF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA552935
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: PIIVIEIYDQ DSMGKADFMG RTFAKPLVKM ADEAYCPPRF PPQLEYYQIY RGNATAGDLL AAFELLQIGP AGKADLPPIN GPVDVDRGPI MPVPMGIRPV LSKYRVEVLF WGLRDLKRVN L Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.
OTOF Polyclonal antibody specifically detects OTOF in Human samples. It is validated for Immunohistochemistry.Spezifikation
OTOF | |
Polyclonal | |
Unconjugated | |
OTOF | |
AUNB1; DFNB6; DFNB9; FER1L2; fer-1-like family member 2; fer-1-like protein 2; NSRD9; Otof; Otoferlin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9381 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HC10 | |
OTOF | |
Recombinant protein corresponding to Human OTOF | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only