Learn More
OTOP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA557333
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: NGVLNESKHQ LNEHKEWLIT LGFGNITTVL DDHTPQCNCT PPTLCTAISH GI Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.
Required for normal formation of otoconia in the inner ear. Inhibits P2Y purinoceptors. Modulates calcium homeostasis and influx of calcium in response to extracellular ATP.Spezifikation
OTOP1 | |
Polyclonal | |
Unconjugated | |
OTOP1 | |
A530025J20Rik; hOtop1; OTOP1; otopetrin 1; otopetrin-1; Otp1; Proton channel OTOP1; tilted; tlt | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
133060 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.30 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7RTM1 | |
OTOP1 | |
Recombinant protein corresponding to Human OTOP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only