missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OTUB2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA551031
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
OTUB2 Polyclonal Antibody detects endogenous levels of OTUB2 protein.
OTUB2 Polyclonal antibody specifically detects OTUB2 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Spezifikation
OTUB2 | |
Polyclonal | |
Unconjugated | |
OTUB2 | |
2010015L18Rik; 4930586I02Rik; AI413508; AW557219; C14orf137; deubiquitinating enzyme OTUB2; FLJ21916; MGC3102; OTB2; OTU deubiquitinase, ubiquitin aldehyde binding 2; OTU domain, ubiquitin aldehyde binding 2; OTU domain-containing ubiquitin aldehyde-binding protein 2; OTU2; OTUB2; Otubain-2; ubiquitin thioesterase OTUB2; ubiquitin-specific protease otubain 2; ubiquitin-specific-processing protease OTUB2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
314405, 68149, 78990 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q96DC9, Q9CQX0 | |
OTUB2 | |
Synthesized peptide derived from the Internal region of human OTUB2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |