missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OTUD4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 25070-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
OTUD4 alternatively spliced transcript variants have been found for this gene. The smaller protein isoform encoded by the shorter transcript variant is found only in HIV-1 infected cells.Spezifikation
OTUD4 | |
Polyclonal | |
Unconjugated | |
OTUD4 | |
DUBA6, HIN1, HIV 1 induced protein HIN 1, HSHIN1, KIAA1046, OTU domain containing 4, OTUD4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54726 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.44 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q01804 | |
OTUD4 | |
OTUD4 Fusion Protein Ag18401 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |