Learn More
OTUD6B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA554964
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: IQGMKNAVPK NDKKRRKQLT EDVAKLEKEM EQKHREELEQ LKLTTKENKI DSVAVNISNL V Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
OTUD6B Polyclonal antibody specifically detects OTUD6B in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
OTUD6B | |
Polyclonal | |
Unconjugated | |
OTUD6B | |
2600013N14Rik; AU015433; CGI-77; Deubiquitinase OTUD6B; DUBA5; DUBA-5; ortholog of human CGI-77; OTU domain containing 6B; OTU domain-containing protein 6B; Otud6b; RGD1310024 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51633 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N6M0 | |
OTUD6B | |
Recombinant protein corresponding to Human OTUD6B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only