missing translation for 'onlineSavingsMsg'
Learn More

Oxalsäure-13C2-Dinatriumsalz, TRC

Artikelnummer. 30419028
Change view
Click to view available options
Menge:
50 mg
Unit Size:
50 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30419028 50 mg 50 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30419028 Supplier Toronto Research Chemicals Supplier No. TRCO84513850MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material Oxalic-13C2 Acid Disodium Salt
CAS 260429-91-2
Summenformel 13C2Na2O4
Menge 50 mg
InChI-Formel InChI=1S/C2H2O4.2Na/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);;/q;2*+1/p-2/i1+1,2+1;;
SMILES O=[13C](O[Na])[13C](O[Na])=O
Molekulargewicht (g/mol) 135.98
Formelmasse 135.9659
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.