missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OXSM Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16642-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a beta-ketoacyl synthetase. The encoded enzyme is required for elongation of fatty acid chains in the mitochondria. Alternatively spliced transcript variants have been described.Spezifikation
OXSM | |
Polyclonal | |
Unconjugated | |
OXSM | |
Beta ketoacyl ACP synthase, FASN2D, KASI, KS, OXSM | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
289934, 54995, 71147 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9D404, Q9NWU1 | |
Oxsm | |
OXSM Fusion Protein Ag10004 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |