Learn More
Oxyphenonium Betaromide, TRC
Artikelnummer. 30414732
Change view
Click to view available options
Menge:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
30414732 50 mg
30411727 100 mg
30417799 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30414732 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCO87696850MG
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 50-10-2
Summenformel C21 H34 N O3 . Br
Molekulargewicht (g/mol) 428.4
Synonym Ethanaminium, 2-[(2-cyclohexyl-2-hydroxy-2-phenylacetyl)oxy]-N,N-diethyl-N-methyl-, bromide (1:1), Ammonium, diethyl(2-hydroxyethyl)methyl-, bromide, α-phenylcyclohexaneglycolate (8CI), Diethyl(2-hydroxyethyl)methylammonium bromide α-phenylcyclohexaneglycolate (6CI), Ethanaminium, 2-[(cyclohexylhydroxyphenylacetyl)oxy]-N,N-diethyl-N-methyl-, bromide (9CI), Cyclohexaneglycolic acid, α-phenyl-, ester with diethyl(2-hydroxyethyl)methylammonium bromide (8CI), (±)-Oxyphenonium bromide, Antrenil, Antrenyl, Antrenyl Duplex, Ba-5473, C 5473, Oxifenon, Oxyfenon, Oxyphenon, Oxyphenonium bromide, Spasmodin, Spasmophen, dl-Oxyphenonium bromide
IUPAC-Name 2-(2-cyclohexyl-2-hydroxy-2-phenylacetyl)oxyethyl-diethyl-methylazanium;bromide
SMILES [Br-].CC[N+](C)(CC)CCOC(=O)C(O)(C1CCCCC1)c2ccccc2
Spezifikation
Chemischer Name oder Material Oxyphenonium Bromide
Menge 50 mg
InChI-Formel InChI=1S/C21H34NO3.BrH/c1-4-22(3,5-2)16-17-25-20(23)21(24,18-12-8-6-9-13-18)19-14-10-7-11-15-19;/h6,8-9,12-13,19,24H,4-5,7,10-11,14-17H2,1-3H3;1H/q+1;/p-1
Formelmasse 427.1722