missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse P-Selectin/CD62P Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 2 publications
N.V.
Spezifikation
Antigen | P-Selectin/CD62P |
---|---|
Klon | 127933 |
Verdünnung | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Anwendungen | Western Blot, ELISA |
Klassifikation | Monoclonal |
Beschreibung
P-Selectin/CD62P Monoclonal specifically detects P-Selectin/CD62P in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Spezifikation
P-Selectin/CD62P | |
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD62P, CD62P antigen, FLJ45155, GMP140, GRMP, PADGEM, PADGEMantigen CD62), PSEL, selectin P (granule membrane protein 140kDa, antigen CD62), SELP | |
SELP | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
127933 | |
Western Blot, ELISA | |
Unconjugated | |
Rat | |
Mouse | |
Q01102 | |
6403 | |
Mouse myeloma cell line NS0-derived recombinant mouse P-Selectin/CD62P Trp42-Ala709 Accession # Q01102 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse P-Selectin/CD62P in ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse (rm) L-Selectin, rmE-Selectin, or recombinant human P-Selectin is observed. |
For Research Use Only