missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ P2Y14 Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 16519801
Change view
Click to view available options
Menge:
100 μL
Packungsgröße:
100 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16519801 100 μL 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 16519801 Lieferant Invitrogen™ Lieferanten-Nr. PA596964

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

Immunogen sequence: ITKKIFKSHL KSSRNSTSVK KKSSRNIFSI VFVFFVCFVP YHIARIPYTK SQTEAHYSCQ SKEILRYMKE FTLLLSAANV CLDPIIYFFL CQPFREILCK KLHIPLKAQN DLDISRIKRG NTTLESTDTL; Positive Samples: Jurkat, U-937, SH-SY5Y, MCF7, Mouse spleen, Mouse lung, Mouse heart, Rat spleen, Rat lung; Cellular Location: Cell membrane, Multi-pass membrane protein.

GPR105 has been reported to be expressed in brain. ESTs have been isolated from kidney, placenta, uterus, and heart/melanocyte/uterus libraries. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more thana thousandmembers currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This turn-off activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Twoblockbuster drugs which have been developed as GPCR-targetedpharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which have multi-billion dollar shares of the mental health and allergy markets, respectively.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen P2Y14
Anwendungen ELISA, Immunohistochemistry (Paraffin), Western Blot
Klassifikation Polyclonal
Konzentration 1.75 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gen P2RY14
Gen-Zugriffsnummer O35881, Q15391, Q9ESG6
Gen-Alias A330108O13Rik; BPR105; G protein coupled receptor for UDP-glucose; G protein-coupled receptor 105; G protein-coupled receptor VTR 15-20; GPCR105; Gpr105; G-protein coupled receptor 105; KIAA0001; P2ry14; P2Y purinoceptor 14; P2Y(14) receptor; P2Y14; P2Y14 receptor; purinergic receptor P2Y, G-protein coupled, 14; purinergic receptor P2Y14; UDP-glucose receptor; VTR 15-20; VTR 15-20 receptor
Gensymbole P2RY14
Wirtsspezies Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 209-338 of human P2RY14 (NP_0556943).
Reinigungsverfahren Affinity Chromatography
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 140795, 171108, 9934
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.