Learn More
P4HA2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA553530
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LKEYILVEEA KLSKIKSWAN KMEALTSKSA ADAEGYLAHP VNAYKLVKRL NTDWPALEDL VLQDSAAGFI ANLSVQRQFF PTDEDEIGAA KALMRLQDTY RLDPGTISRG ELPGTKYQAM LSVDDCFGM Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 91%.
P4HA2 Polyclonal antibody specifically detects P4HA2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
P4HA2 | |
Polyclonal | |
Unconjugated | |
P4HA2 | |
4-PH alpha 2; 4-PH alpha-2; 4PHalpha2; AA407196; C76437; collagen prolyl 4-hydroxylase alpha(II); C-P4Halpha(II); hypothetical protein LOC507327; P4HA2; P4hl; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha II polypeptide; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide 2; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide II; procollagen-proline,2-oxoglutarate-4-dioxygenase subunit alpha-2; prolyl 4-hydroxylase A; Prolyl 4-hydroxylase alpha IIa subunit; prolyl 4-hydroxylase subunit alpha 2; prolyl 4-hydroxylase subunit alpha-2; prolyl 4-hydroxylase, alpha polypeptide II; UNQ290/PRO330; zgc:91948 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8974 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15460 | |
P4HA2 | |
Recombinant protein corresponding to Human P4HA2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only