missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PABPC4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 14960-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Protein phosphatase 2 is one of the four major Ser/Thr phosphatases and is implicated in the negative control of cell growth and division. Protein phosphatase 2 holoenzymes are heterotrimeric proteins composed of a structural subunit A, a catalytic subunit C, and a regulatory subunit B. The regulatory subunit is encoded by a diverse set of genes that have been grouped into the B/PR55, B′/PR61, and B′′/PR72 families. These different regulatory subunits confer distinct enzymatic specificities and intracellular localizations to the holozenzyme. The product of this gene belongs to the B′′ family. The B′′ family has been further divided into subfamilies. The product of this gene belongs to the beta subfamily of regulatory subunit B′′. Alternative splicing results in multiple transcript variants encoding different isoforms.Spezifikation
PABPC4 | |
Polyclonal | |
Unconjugated | |
PABPC4 | |
Activated platelet protein 1, APP 1, APP1, iPABP, PABP 4, PABP4, PABPC4, Poly(A) binding protein 4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
230721, 298510, 8761 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunocytochemistry, Immunofluorescence | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13310 | |
PABPC4 | |
PABPC4 Fusion Protein Ag6813 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |