missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PABPC4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 € - 399.00 €
Spezifikation
Antigen | PABPC4 |
---|---|
Konzentration | 0.33 mg/mL |
Anwendungen | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16883894
|
Proteintech
14960-1-AP-20UL |
20 μL |
140.00 €
20 Mikroliter |
Verfügbar ab: 25-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16873894
|
Proteintech
14960-1-AP-150UL |
150 μL |
399.00 €
150 Mikroliter |
Verfügbar ab: 25-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Protein phosphatase 2 is one of the four major Ser/Thr phosphatases and is implicated in the negative control of cell growth and division. Protein phosphatase 2 holoenzymes are heterotrimeric proteins composed of a structural subunit A, a catalytic subunit C, and a regulatory subunit B. The regulatory subunit is encoded by a diverse set of genes that have been grouped into the B/PR55, B′/PR61, and B′′/PR72 families. These different regulatory subunits confer distinct enzymatic specificities and intracellular localizations to the holozenzyme. The product of this gene belongs to the B′′ family. The B′′ family has been further divided into subfamilies. The product of this gene belongs to the beta subfamily of regulatory subunit B′′. Alternative splicing results in multiple transcript variants encoding different isoforms.Spezifikation
PABPC4 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q13310 | |
230721, 298510, 8761 | |
PABPC4 Fusion Protein Ag6813 | |
Primary | |
-20°C | |
PABPC4 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Activated platelet protein 1, APP 1, APP1, iPABP, PABP 4, PABP4, PABPC4, Poly(A) binding protein 4 | |
PABPC4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |