missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PAK7 (KIAA1264), His Tag Recombinant Protein

Artikelnummer. 10258302
Change view
Click to view available options
Menge:
1 mg
Packungsgröße:
1 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
10258302 1 mg 1 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.

Artikelnummer. 10258302 Lieferant Invitrogen™ Lieferanten-Nr. PV4441

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Catalytic.

PAK7 (KIAA1264) is implicated in the regulation of cytoskeletal dynamics, proliferation, and cell survival signaling. This kinase contains a CDC42/Rac1 interactive binding motif, and has been shown to bind CDC42 in the presence of GTP. This kinase is predominantly expressed in brain. It is capable of promoting neurite outgrowth, and thus may play a role in neurite development. This kinase is associated with microtubule networks and induces microtubule stabilization. The subcellular localization of this kinase is tightly regulated during cell cycle progression. Alternatively spliced transcript variants encoding the same protein have been described.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer Q9P286
Konzentration See Label
Zur Verwendung mit (Anwendung) Kinase Assay
Zusammensetzung 50mM tris with 0.02% Triton X-100, 2mM DTT, 150mM NaCl, 0.5mM EDTA, 50% glycerol and no preservative; pH 7.5
Gen-ID (Entrez) 57144
Molekulargewicht 37.6 kDa
Name Human PAK7 (KIAA1264), His Tag
pH-Bereich 7.5
Reinigungsverfahren Purified
Menge 1 mg
Lagerungsbedingungen -80°C, Avoid Freeze/Thaw Cycles
Kennzeichnung RUO
Gen-Alias 2900083L08Rik; 6430627N20; KIAA1264; p21 (CDKN1A)-activated kinase 7; p21 (RAC1) activated kinase 5; p21 (RAC1) activated kinase 7; p21 protein (Cdc42/Rac)-activated kinase 7; p21(CDKN1A)-activated kinase 7; p21-activated kinase 5; p21-activated kinase 7; p21CDKN1A-activated kinase 7; PAK 5; PAK 7; PAK5; PAK-5; Pak7; PAK-7; protein kinase PAK5; RP5-1119D9.3; Serine/threonine-protein kinase PAK 5; serine/threonine-protein kinase PAK 7; serine/threonine-protein kinase PAK7
Gebräuchliche Bezeichnung PAK7
Gensymbol PAK5
Kinase-Familie STE20 Kinase Family
Proteinlänge 425-719
Produkttyp Protein
Konjugat Unconjugated
Spezies Human
Rekombinant Recombinant
Proteinmarkierung His-tag
Sequenz 425-719
Inhalt und Lagerung -80°C, Avoid Freeze/Thaw Cycles
Kinase-Gruppe Ser/Thr Kinases: STE Group
Versandbedingung Dry Ice
Expressionssystem Baculovirus
Proteinfamilie Kinases & Inhibitors
Proteinform Catalytic
Form Liquid
Forschungskategorie Drug Development
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.