Learn More
Palmitoylethanolamide, Tocris Bioscience™
Selective GPR55 agonist. FAAH and PAA substrate
Marke:  Tocris (Bio-Techne) 087950
Weitere Details : Netto-Gewicht : 0.01000kg
Menge. 50mg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13418103
  • 469.32 € / 50 Milligramm
Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Endogenous lipid that acts as a selective GPR55 agonist (EC50 values are 4, 19 800 and > 30 000 nM at GPR55, CB2 and CB1 receptors respectively). Substrate for fatty acid amide hydrolase (FAAH) and PEA-preferring acid amidase (PAA) and exhibits antinociceptive and anticonvulsant in vivo. Directly activates PPARα (EC50 = 3 μM) producing robust anti-inflammatory actions.
Spezifikation
Chemische Identifikatoren
544-31-0
299.499
palmitoylethanolamide, palmidrol, n-2-hydroxyethyl hexadecanamide, palmitamide mea, hydroxyethylpalmitamide, n-palmitoylethanolamine, n-2-hydroxyethyl palmitamide, impulsin, palmitic acid monoethanolamide, loramine p 256
CHEBI:71464
CCCCCCCCCCCCCCCC(=O)NCCO
C18H37NO2
HXYVTAGFYLMHSO-UHFFFAOYSA-N
4671
N-(2-hydroxyethyl)hexadecanamide
Spezifikation
Palmitoylethanolamide
544-31-0
palmitoylethanolamide, palmidrol, n-2-hydroxyethyl hexadecanamide, palmitamide mea, hydroxyethylpalmitamide, n-palmitoylethanolamine, n-2-hydroxyethyl palmitamide, impulsin, palmitic acid monoethanolamide, loramine p 256
CCCCCCCCCCCCCCCC(=O)NCCO
299.499
CHEBI:71464
>98%
50mg
C18H37NO2
HXYVTAGFYLMHSO-UHFFFAOYSA-N
N-(2-hydroxyethyl)hexadecanamide
4671
299.5
Videos