Learn More
Pantoprazole sodium hydrate, MedChemExpress
Product Code. 16467346
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16467346 100 mg 100mg
16421876 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16467346 Supplier Medchem Express Supplier No. HY17507B
 Restricted Item
Verfügbar ab: 01-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MedChemExpress Pantoprazole sodium hydrate (BY10232 sodium hydrate) is an orally active and potent proton pump inhibitor (PPI).
Pantoprazole sodium hydrate, a substituted benzimidazole, is a potent H+/K+-ATPase inhibitor with an IC50 of 6.8 μM. Pantoprazole sodium hydrate improves pH stability and has anti-secretory, anti-ulcer activities. Pantoprazole sodium hydrate significantly increased tumor growth delay combined with Doxorubicin (HY-15142).
Specifications
Chemical Name or Material Pantoprazole sodium hydrate
CAS 164579-32-2
Color White
Purity Grade Notes Research
Molecular Formula C16H17F2N3NaO5.5S
Quantity 100 mg
Synonym BY1023 sodium hydrateSKF96022 sodium hydrate
Solubility Information H2O : 250 mg/mL (578.21 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (231.28 mM; Need ultrasonic)
SMILES O=S(C1=NC2=CC=C(OC(F)F)C=C2N1[Na])CC3=NC=CC(OC)=C3OC.[1.5H2O]
Molecular Weight (g/mol) 432.37
Formula Weight 432.37
Percent Purity 99.85%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neurodegeneration