missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAX5 / BSAP (Early B-Cell Marker) Mouse, Clone: PCRP-PAX5-1B7, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio 5079-MSM1-P1
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
PAX5 / BSAP (Early B-Cell Marker) | |
Nucleus | |
PCRP-PAX5-1B7 | |
Unconjugated | |
Mouse | |
42kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
5079 | |
IgG1 |
Immunoprecipitation, Western Blot, Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant full-length human PAX5 protein | |
Raji, DAUDI or Ramos Cells. Tonsil, spleen, Lymph Node or Colon Carcinoma. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |