missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCDHB5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15245-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Potential calcium-dependent cell-adhesion protein. May be involved in the establishment and maintece of specific neuronal connections in the brain.Spezifikation
PCDHB5 | |
Polyclonal | |
Unconjugated | |
PCDHB5 | |
PCDH beta 5, PCDH BETA5, PCDHB5, protocadherin beta 5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26167, 93876 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9Y5E4 | |
Pcdhb5 | |
PCDHB5 Fusion Protein Ag7458 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |