Beschreibung
PD-1 Monoclonal antibody specifically detects PD-1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Spezifikation
Spezifikation
| Antigen | PD-1 |
| Anwendungen | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Klon | CL3624 |
| Konjugat | mFluor Violet 500 SE |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 |
| Wirtsspezies | Mouse |
| Immunogen | This antibody was developed using a recombinant protein derived from Q15116, with the exact immunogen sequence remaining proprietary. |
| Reinigungsverfahren | Protein A purified |
| Mehr anzeigen |