missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentafluorphenyl2-pyrid-3-yl-1,3-thiazol-4-carboxylat, 97 %, Thermo Scientific™
Click to view available options
Menge:
250 mg
1 g
Spezifikation
Chemischer Name oder Material | Pentafluorophenyl 2-pyrid-3-yl-1, 3-thiazole-4-carboxylate |
CAS Min. % | 96.5 |
CAS Max. % | 100.0 |
Prozentgehaltsbereich | 97% |
Menge | 250 mg |
IUPAC-Name | 2,3,4,5,6-Pentafluorphenyl-2-(pyridin-3-yl)-1,3-thiazol-4-carboxylat |
Formelmasse | 372.27 |
Reinheit (%) | 97% |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'