missing translation for 'onlineSavingsMsg'
Learn More

Perfluor-1-Ioddecan, 97 %, Thermo Scientific Chemicals

Catalog Number p-7026061
Click to view available options
Menge:
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
423-62-1
C10F21I
645.981
MFCD00001065
UDWBMXSQHOHKOI-UHFFFAOYSA-N
perfluorodecyl iodide, 1-iodoperfluorodecane, perfluoro-1-iododecane, henicosafluoro-10-iododecane, heneicosafluorodecyl iodide, heneicosafluoro-1-iododecane, decane, heneicosafluoro-1-iodo, iodoperfluorodecane, henicosafluorodecyl iodide, 1-iodohenicosafluorodecane
67920
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-Henicosafluoro-10-iododecan
C(C(C(C(C(C(F)(F)I)(F)F)(F)F)(F)F)(F)F)(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
423-62-1
C10F21I
645.981
MFCD00001065
UDWBMXSQHOHKOI-UHFFFAOYSA-N
perfluorodecyl iodide, 1-iodoperfluorodecane, perfluoro-1-iododecane, henicosafluoro-10-iododecane, heneicosafluorodecyl iodide, heneicosafluoro-1-iododecane, decane, heneicosafluoro-1-iodo, iodoperfluorodecane, henicosafluorodecyl iodide, 1-iodohenicosafluorodecane
67920
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-Henicosafluoro-10-iododecan
C(C(C(C(C(C(F)(F)I)(F)F)(F)F)(F)F)(F)F)(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Perfluoro-1-iododecane
Schmelzpunkt 63°C to 67°C
Siedepunkt 195°C to 200°C
Flammpunkt None
Menge 5 g
Beilstein 1810506
Empfindlichkeit Light sensitive
Formelmasse 645.98
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'