Learn More
Alfa Aesar™ Perfluordodecan, 97 %
Marke: Alfa Aesar™ L16827.03
Weitere Details : CAS : 307-59-5 Netto-Gewicht : 0.00100kg
Menge | 1g |
---|
Chemische Identifikatoren
307-59-5 | |
638.09 | |
WNZGTRLARPEMIG-UHFFFAOYSA-N | |
78973 | |
C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F |
C12F26 | |
MFCD00042084 | |
perfluorododecane, hexacosafluorododecane, n-perfluorododecane, perfluoro-n-dodecane, acmc-20ajy2 | |
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-hexacosafluorododecan |
Spezifikation
Perfluorododecane | |
178°C | |
1g | |
C12F26 | |
MFCD00042084 | |
perfluorododecane, hexacosafluorododecane, n-perfluorododecane, perfluoro-n-dodecane, acmc-20ajy2 | |
C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F | |
638.09 | |
638.09 |
1.5 | |
72°C to 76°C | |
307-59-5 | |
CF3(CF2)10CF3 | |
1811139 | |
WNZGTRLARPEMIG-UHFFFAOYSA-N | |
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-hexacosafluorododecan | |
78973 | |
97% |