missing translation for 'onlineSavingsMsg'
Learn More

peroxiredoxin 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 10545-2-AP-150UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16886993

  • 399.00 € / 150 Mikroliter
Verfügbar ab: 04-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This gene encodes a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. The encoded protein may play an antioxidant protective role in cells, and may contribute to the antiviral activity of CD8 (+) T-cells. This protein may have a proliferative effect and play a role in cancer development or progression. The crystal structure of this protein has been resolved to 2. 7 angstroms. Transcript variants encoding distinct isoforms have been identified for this gene.
Spezifikation

Spezifikation

peroxiredoxin 2
Polyclonal
Unconjugated
PRDX2
NKEF B, NKEFB, peroxiredoxin 2, PRDX2, PRP, PRX II, PRX2, PRXII, TDPX1, Thioredoxin peroxidase 1, TSA
Rabbit
Antigen Affinity Chromatography
RUO
21672, 29338, 7001
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P32119, P35704, Q61171
PRDX2
peroxiredoxin 2 Fusion Protein Ag0835
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote