missing translation for 'onlineSavingsMsg'
Learn More

PF 429242, Tocris Bioscience™

Competitive inhibitor of SREBP site 1 protease

Marke:  RD Systems (Bio-Techne) 3354/50

Weitere Details : Netto-Gewicht : 0.00050kg

Menge. 50mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412874

  • 1209.54 € / 50 Milligramm


Voraussichtlicher Liefertermin 30-04-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Reversible, competitive inhibitor of sterol regulatory element-binding protein (SREBP) site 1 protease (IC 50 = 0.175 μM). Selective for site 1 protease against a panel of serine proteases. Inhibits rate of cholesterol synthesis in CHO cells (IC 50 = 0.53 μM). Also displays antiviral activity. Cell permeable.
Spezifikation

Chemische Identifikatoren

947303-87-9
409.574
r-4-diethylamino methyl-n-2-methoxyphenethyl-n-pyrrolidin-3-yl benzamide, unii-49wb3oa7vn, 49wb3oa7vn, r-4-diethylamino methyl-n-2-methoxyphenethyl-n-pyrrolidin-3-yl benz amide, 4-diethylamino methyl-n-2-2-methoxyphenyl ethyl-n-3r-3-pyrrolidinylbenzamide, 4-diethylamino methyl-n-2-2-methoxyphenyl ethyl-n-3r-pyrrolidin-3-yl benzamide, d0p7ny, benzamide, 4-diethylamino methyl-n-2-2-methoxyphenyl ethyl-n-3r-3-pyrrolidinyl, 4-diethylamino methyl-n-2-2-methoxyphenyl eth, r-n-2-methoxyphenethyl-4-diethylamino methyl-n-pyrrolidin-3-yl benzamide
4-(diethylaminomethyl)-N-[2-(2-methoxyphenyl)ethyl]-N-[(3R)-pyrrolidin-3-yl]benzamide
C25H35N3O2
XKPJTOHUPQWSOJ-HSZRJFAPSA-N
23661637
CCN(CC)CC1=CC=C(C=C1)C(=O)N(CCC2=CC=CC=C2OC)C3CCNC3

Spezifikation

PF 429242
947303-87-9
r-4-diethylamino methyl-n-2-methoxyphenethyl-n-pyrrolidin-3-yl benzamide, unii-49wb3oa7vn, 49wb3oa7vn, r-4-diethylamino methyl-n-2-methoxyphenethyl-n-pyrrolidin-3-yl benz amide, 4-diethylamino methyl-n-2-2-methoxyphenyl ethyl-n-3r-3-pyrrolidinylbenzamide, 4-diethylamino methyl-n-2-2-methoxyphenyl ethyl-n-3r-pyrrolidin-3-yl benzamide, d0p7ny, benzamide, 4-diethylamino methyl-n-2-2-methoxyphenyl ethyl-n-3r-3-pyrrolidinyl, 4-diethylamino methyl-n-2-2-methoxyphenyl eth, r-n-2-methoxyphenethyl-4-diethylamino methyl-n-pyrrolidin-3-yl benzamide
CCN(CC)CC1=CC=C(C=C1)C(=O)N(CCC2=CC=CC=C2OC)C3CCNC3
409.574
482.49
50mg
C25H35N3O2
XKPJTOHUPQWSOJ-HSZRJFAPSA-N
4-(diethylaminomethyl)-N-[2-(2-methoxyphenyl)ethyl]-N-[(3R)-pyrrolidin-3-yl]benzamide
23661637
>97%
Videos