missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFDN5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15078-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Binds specifically to cytosolic chaperonin (c-CPN) and transfers target proteins to it. Binds to nascent polypeptide chain and promotes folding in an environment in which there are many competing pathways for nonnative proteins. Represses the transcriptional activity of MYC.Spezifikation
PFDN5 | |
Polyclonal | |
Unconjugated | |
PFDN5 | |
C Myc binding protein Mm 1, MM 1, MM1, Myc modulator 1, PFD5, PFDN5, prefoldin subunit 5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
300257, 5204, 56612 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunocytochemistry | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99471, Q9WU28 | |
PFDN5 | |
PFDN5 Fusion Protein Ag7150 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |