missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGAM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16126-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This antibody is specific to PGAM1.
PGAM1 belongs to the phosphoglycerate mutase family. PGAM1 is an important component of glucose and 2,3-BPGA (2,3-bisphosphoglycerate) metabolism and catalyzes the reversible reaction of 3-phosphoglycerate (3-PGA) to 2-phosphoglycerate (2-PGA) in the glycolytic pathway. The PGAM1 protein is a dimeric enzyme containing, in different tissues, different proportions of a slow-migrating muscle (MM) isozyme, a fast-migrating brain (BB) isozyme, and a hybrid form (MB). Mutations in PGAM1 cause muscle phosphoglycerate mutase efficiency, also known as glycogen storage disease X. Recombit human PGAM protein, fused to His-tag at N-terminus, was expressed in E.coli and purified by using conventional chromatography techniques.Spezifikation
PGAM1 | |
Polyclonal | |
Unconjugated | |
PGAM1 | |
BPG dependent PGAM 1, PGAM B, PGAM1, PGAMA, Phosphoglycerate mutase 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18648, 24642, 5223 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P18669, P25113, Q9DBJ1 | |
PGAM1 | |
PGAM1 Fusion Protein Ag9110 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |