Learn More
Phenoxybenzamine hydrochloride, MedChemExpress
Product Code. 16452169
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
500 mg
1 g
Unit Size:
1g
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452169 1 g 1g
16461416 200 mg 200mg
16422136 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16452169 Supplier Medchem Express Supplier No. HYB0431A
 Restricted Item
Verfügbar ab: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MedChemExpress Phenoxybenzamine hydrochloride is a nonselective, irreversible, orally active α-adrenoceptor antagonist that is commonly used for the research of hypertension, specifically caused by pheochromocytoma. Phenoxybenzamine hydrochloride also shows antitumor activity.
Phenoxybenzamine hydrochloride is a nonselective, irreversible, orally active α-adrenoceptor antagonist that is commonly used for the research of hypertension, specifically caused by pheochromocytoma. Phenoxybenzamine hydrochloride also shows antitumor activity.
Specifications
Chemical Name or Material Phenoxybenzamine hydrochloride
CAS 63-92-3
Color White
Purity Grade Notes Research
Molecular Formula C18H23Cl2NO
Quantity 1 g
Solubility Information DMSO : 100 mg/mL (293.87 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (1.97 mM; Need ultrasonic)
SMILES CC(COC1=CC=CC=C1)N(CCCl)CC2=CC=CC=C2.Cl
Molecular Weight (g/mol) 340.29
Formula Weight 340.29
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death