missing translation for 'onlineSavingsMsg'
Learn More

Phenyl isocyanate, 98+%, Thermo Scientific™

Product Code. 11481910
Click to view available options
Quantity:
5 g
250 g
1000 g
Unit Size:
1000g
250g
5g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 11481910

Brand: Thermo Scientific Alfa Aesar A12808.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 618 41 11 for assistance
View alternative products

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Phenyl isocyanate is used with triethylamine in order to activate nitro groups to undergo 1,3-dipolar cycloaddition. It is also employed in the preparation of chemically modified cellulose paper. Further, it is used in the preparation of functionalized graphene oxide nanoplatelets. In addition to this, it reacts with ethyl alcohol to prepare ethyl alfa,gamma-diphenylallophanate and small amount of phenyl isocyanate dimer.

Solubility
Miscible with ether.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, water, moisture, alcohols, amines, strong bases and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-71-9
Molecular Formula C7H5NO
Molecular Weight (g/mol) 119.123
MDL Number MFCD00001994
InChI Key DGTNSSLYPYDJGL-UHFFFAOYSA-N
Synonym phenyl isocyanate, phenylcarbimide, carbanil, phenyl carbonimide, benzene, isocyanato, phenylisocyanate, isocyanic acid, phenyl ester, mondur p, phnco, fenylisokyanat
PubChem CID 7672
ChEBI CHEBI:53806
IUPAC Name isocyanatobenzene
SMILES C1=CC=C(C=C1)N=C=O

Specifications

Melting Point -33°C
Density 1.094
Boiling Point 62°C to 64°C (11 mmHg)
Flash Point 51°C (123°F)
Odor Pungent
Refractive Index 1.536
Quantity 1000 g
UN Number UN2487
Beilstein 471391
Sensitivity Moisture sensitive
Merck Index 14,7296
Solubility Information Miscible with ether.
Formula Weight 119.12
Percent Purity ≥98%
Chemical Name or Material Phenyl isocyanate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.