missing translation for 'onlineSavingsMsg'
Learn More

(Phenylthio)essigsäure, 97 %, Thermo Scientific Chemicals

Catalog Number p-7026329
Click to view available options
Menge:
25 g
100 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
103-04-8
C8H7O2S
167.20
MFCD00004355
MOTOSAGBNXXRRE-UHFFFAOYSA-M
phenylthio acetic acid, thiophenoxyacetic acid, 2-phenylthio acetic acid, phenylthioglycolic acid, phenylmercaptoacetic acid, 2-phenylsulfanyl acetic acid, phenylsulfanyl acetic acid, acetic acid, phenylthio, phenylmercapto acetic acid, carboxymethyl phenyl sulfide
59541
2-(phenylsulfanyl)acetate
[O-]C(=O)CSC1=CC=CC=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
103-04-8
C8H7O2S
167.20
MFCD00004355
MOTOSAGBNXXRRE-UHFFFAOYSA-M
phenylthio acetic acid, thiophenoxyacetic acid, 2-phenylthio acetic acid, phenylthioglycolic acid, phenylmercaptoacetic acid, 2-phenylsulfanyl acetic acid, phenylsulfanyl acetic acid, acetic acid, phenylthio, phenylmercapto acetic acid, carboxymethyl phenyl sulfide
59541
2-(phenylsulfanyl)acetate
[O-]C(=O)CSC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material (Phenylthio)acetic acid
Schmelzpunkt 61°C to 66°C
Geruch Stench
Menge 25 g
Beilstein 2045072
Formelmasse 168.22
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'