missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHKA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 24279-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Phosphorylase b kinase catalyzes the phosphorylation of serine in certain substrates, including troponin I. The alpha chain may bind calmodulin.Spezifikation
PHKA1 | |
Polyclonal | |
Unconjugated | |
PHKA1 | |
PHKA, PHKA1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18679, 5255, 64561 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P18826, P46020, Q64649 | |
Phka1 | |
PHKA1 Fusion Protein Ag17998 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |