Learn More
PHKA2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA551772
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: GARVKLGNLS EFLTTSFYTY LTFLDPDCDE KLFDNASEGT FSPDSDSDLV GYLEDTCNQE SQDELDHYIN HLLQSTSLRS YLPPLCKNTE DRHVFSAIHS TRDILSVMAK AKGLEVPFVP MTLPTKVLSA HRKS Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 36%.
PHKA2 Polyclonal antibody specifically detects PHKA2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
PHKA2 | |
Polyclonal | |
Unconjugated | |
PHKA2 | |
6330505C01Rik; D330034O08; GSD9A; Phk; Phka2; PHKLA; phosphorylase b kinase regulatory subunit alpha liver isoform; phosphorylase b kinase regulatory subunit alpha, liver isoform; phosphorylase kinase; phosphorylase kinase alpha 2; phosphorylase kinase alpha L subunit; phosphorylase kinase alpha-subunit; phosphorylase kinase regulatory subunit alpha 2; phosphorylase kinase, alpha 2; phosphorylase kinase, alpha 2 (liver); PYK; PYKL; RP3-499B10.2; XLG; XLG2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5256 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P46019 | |
PHKA2 | |
Recombinant protein corresponding to Human PHKA2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only