missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-HDAC5 (Ser259) Polyclonal Antibody
GREENER_CHOICE

Product Code. 16671485
Change view
Click to view available options
Menge:
100 μL
Unit Size:
100 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
16671485 100 μL 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16671485 Supplier Invitrogen™ Supplier No. PA5105040

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of HDAC5 only when phosphorylated at Serine 259.

Histones play a critical role in transcriptional regulation, cell cycle progression, and developmental events. Histone acetylation/deacetylation alters chromosome structure and affects transcription factor access to DNA. The protein encoded by this gene belongs to the class II histone deacetylase/acuc/apha family. It possesses histone deacetylase activity and represses transcription when tethered to a promoter. It coimmunoprecipitates only with HDAC3 family member and might form multicomplex proteins. It also interacts with myocyte enhancer factor-2 (MEF2) proteins, resulting in repression of MEF2-dependent genes. This gene is thought to be associated with colon cancer. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-HDAC5 (Ser259)
Anwendungen Western Blot, Immunocytochemistry
Klassifikation Polyclonal
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gen HDAC5
Gen-Zugriffsnummer Q9UQL6, Q9Z2V6
Gen-Alias AI426555; Antigen NY-CO-9; DKFZp779K1053; FLJ90614; HD5; HD7; HD7b; HD9; HDAC; Hdac4; HDAC5; HDAC-5; HDAC7; HDAC7B; HDAC9; HDAC9B; HDAC9FL; HDRP; histone deacetylase 4; histone deacetylase 5; histone deacetylase mHDA1; KIAA0600; KIAA0744; mHDA1; mKIAA0600; NY-CO-9
Gensymbole HDAC5
Wirtsspezies Rabbit
Immunogen A synthesized peptide derived from human HDAC5(Accession Q9UQL6), corresponding to amino acid residues around phosphorylated Ser259.
Reinigungsverfahren Sequential chromatography
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 10014, 15184
Zielspezies Human, Mouse
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.