missing translation for 'onlineSavingsMsg'
Learn More

Phospho-p21 (Thr57) Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA564842

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 15742906

  • 471.00 € / 100 Mikroliter
Verfügbar ab: 19-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Phospho-p21 (Thr57) Polyclonal Antibody detects endogenous levels of p21 only when phosphorylated at Thr57.

Phospho-p21 (Thr57) Polyclonal antibody specifically detects Phospho-p21 (Thr57) in Human, Mouse, Rat samples. It is validated for Western Blot.
Spezifikation

Spezifikation

Phospho-p21 (Thr57)
Polyclonal
Unconjugated
CDKN1A
Activating Fragment 1; CAP20; cdk inhibitor CIP1; cdk inhibitor CIP1 (p21); CDKI; CDK-interacting protein 1; CDK-interaction protein 1; Cdkn1; CDKN1A; CDNK1A; CIP1; Cip1); cyclin dependent kinase inhibitor 1A; cyclin-dependent kinase inhibitor 1; cyclin-dependent kinase inhibitor 1A; Cyclin-dependent kinase inhibitor 1A (p21; cyclin-dependent kinase inhibitor 1A (P21); cyclin-dependent kinase inhibitor 1A (p21, Cip1); cyclin-dependent kinase inhibitor 1A variant 1; cyclin-dependent kinase inhibitor 1A variant 2; DNA synthesis inhibitor; MDA6; MDA-6; melanoma differentiation associated protein 6; melanoma differentiation-associated protein; Melanoma differentiation-associated protein 6; OTTHUMP00000016298; p21; p21<WAF1/CIP1>; p21Cip1; p21Cip1/Waf1; p21WAF; PIC1; putative cyclin-dependent kinase inhibitor 1A (p21/WAF1/CIP1); SDI1; SLC12A9; UV96; WAF1; Wild type p53 activated fragment 1 (WAF1); wild-type p53-activated fragment 1
Rabbit
Affinity chromatography
RUO
1026, 114851, 12575
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P38936, P39689
CDKN1A
A synthesized peptide derived from human p21Cip1 (Phospho-Thr57)
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote