missing translation for 'onlineSavingsMsg'
Learn More

Phospho-Syk (Tyr323, Tyr317) Recombinant Polyclonal Antibody (6HCLC), Invitrogen™

Rabbit Recombinant Polyclonal Antibody

Marke:  Invitrogen 711415

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 15647798

  • 461.00 € / 100 Mikrogramm
Verfügbar ab: 08-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.


Beschreibung

Beschreibung

This antibody is predicted to react with Monkey, Cat, Rabbit and Horse Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.

Phospho-Syk (Tyr323, Tyr317) Recombinant Polyclonal antibody specifically detects Phospho-Syk (Tyr323, Tyr317) in Human samples. It is validated for Immunocytochemistry, Western Blot.
Spezifikation

Spezifikation

Phospho-Syk (Tyr323, Tyr317)
Recombinant Polyclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P43405
SYK
Peptide corresponding to Human SYK (aa 319-327)
100 μg
Primary
Human
Antibody
IgG
Immunocytochemistry, Western Blot
6HCLC
Unconjugated
SYK
DKFZp313N1010; EC 2.7.10.2; FLJ25043; FLJ37489; HGNC:11205; kinase Syk; KSYK; p72Syk; p72-Syk; ptk72; SP1; spleen associated tyrosine kinase; spleen tyrosine kinase; SYK; syk signaling; Sykb; tyrosine-kinase syk; tyrosine-protein kinase SYK
Rabbit
Protein A
RUO
6850
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote