missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-TAB1 (Ser438) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA564720
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Phospho-TAB1 (Ser438) Polyclonal Antibody detects endogenous levels of TAB1 only when phosphorylated at Ser438.
Phospho-TAB1 (Ser438) Polyclonal antibody specifically detects Phospho-TAB1 (Ser438) in Human, Mouse samples. It is validated for Western Blot.Spezifikation
Phospho-TAB1 (Ser438) | |
Polyclonal | |
Unconjugated | |
TAB1 | |
2310012M03Rik; 3'-Tab1; b2b449Clo; beta activated kinase-1 binding protein-1; MAP3K7IP1; MGC57664; mitogen activated protein kinase kinase kinase 7 interacting protein 1; mitogen-activated protein kinase kinase kinase 7 interacting protein 1; mitogen-activated protein kinase kinase kinase 7-interacting protein 1; RP3-407F17.2; TAB1; TAB1beta; TAK1-binding protein 1; TGF-beta activated kinase 1/MAP3K7 binding protein 1; TGF-beta-activated kinase 1 and MAP3K7-binding protein 1; TGF-beta-activated kinase 1-binding protein 1; transforming growth factor beta-activated kinase-binding protein 1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
10454, 66513 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 30% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15750, Q8CF89 | |
TAB1 | |
A synthesized peptide derived from human TAB1 (Phospho-Ser438) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |