missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-XRCC4 (Ser260) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA564731
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Phospho-XRCC4 (Ser260) Polyclonal Antibody detects endogenous levels of XRCC4 only when phosphorylated at Ser260.
Phospho-XRCC4 (Ser260) Polyclonal antibody specifically detects Phospho-XRCC4 (Ser260) in Human samples. It is validated for Western Blot.Spezifikation
Phospho-XRCC4 (Ser260) | |
Polyclonal | |
Unconjugated | |
XRCC4 | |
2310057B22Rik; AW413319; AW545101; DNA repair protein XRCC4; SSMED; X-ray repair complementing defective repair in Chinese hamster cells 4; X-ray repair cross complementing 4; X-ray repair cross complementing protein 4; X-ray repair cross-complementing protein 4; XRCC4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
7518 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q13426 | |
XRCC4 | |
A synthesized peptide derived from human XRCC4 (Phospho-Ser260) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |